missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Parts/Accessories for OmniPAGE Mini Vertical Electrophoresis System

Product Code. 11873293 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
OmniPAGE Mini CVS10 Vertical System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11873293 OmniPAGE Mini CVS10 Vertical System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11873293
Only null left
Add to basket
Add to basket

External casting system (upstand and base), spare tank or spare lid. Fisherbrand™ Parts/Accessories for OmniPAGE Mini Vertical Electrophoresis System are used for applications involving the separation of proteins and nuclueic acids in poly-acrilamide gels.

OmniPAGE Mini Vertical systems are used primarily for protein but also nucleic acid electrophoresis techniques. By combining both functionality and ease of use, the OmniPAGE CVS10 systems enable simple, versatile vertical mini gel electrophoresis with reproducible results.

Each OmniPAGE system can accommodate up to 4 hand-cast gels and 2 commercial precast gels to provide complete flexibility. Unique sliding clamp technology within the PAGE insert facilitates fast, intuitive leak-free casting.

  • Electrophoresis Tank: Holds the gel module with cast gels/samples and the buffer solution; connects to an external power source
  • Safety Lid: Ensures the correct polarity and orientation of electrodes; fits easily onto tank
  • External Casting System: includes upstand and base with silicone mat; securely holds gel module while casting gel
All parts may be used with glass plates/PAGE insert, blotting insert or tube gel insert.

Specifications

Color White
For Use With (Application) Protein Gel Electrophoresis
For Use With (Equipment) OmniPAGE Mini CVS10 Vertical System
Type External Casting System
Gel Dimensions 8 x 8.5 cm
Includes Upstand and base
No. of Gels Up to 2
Dimensions (L x W) 10 x 10 cm (plate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.