missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEFT Mouse anti-Human, Mouse, Rat, Clone: 5A11F8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66012-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).Specifications
GEFT | |
Monoclonal | |
1.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P720, Q86VW2, Q9CWR0 | |
Arhgef25 | |
GEFT Fusion Protein Ag10819 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
5A11F8 | |
Unconjugated | |
ARHGEF25 | |
ARHGEF25, GEFT, p63RhoGEF, RhoA/RAC/CDC42 exchange factor | |
Mouse | |
Protein A | |
RUO | |
115557, 314904, 52666 | |
-20°C | |
Liquid |