missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEFT Mouse anti-Human, Mouse, Rat, Clone: 5A11F8, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | GEFT |
---|---|
Clone | 5A11F8 |
Concentration | 1.34 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877505
|
Proteintech
66012-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867505
|
Proteintech
66012-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).Specifications
GEFT | |
1.34 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARHGEF25, GEFT, p63RhoGEF, RhoA/RAC/CDC42 exchange factor | |
Arhgef25 | |
IgG2a | |
Protein A | |
Antibody |
5A11F8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human, Rat | |
Q6P720, Q86VW2, Q9CWR0 | |
115557, 314904, 52666 | |
GEFT Fusion Protein Ag10819 | |
Primary | |
-20°C | |
ARHGEF25 |