missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Maximum resolution achieved from efficiency of small spherical particles, optimally packed
- Particles are coupled to Q or S ion exchangers with excellent sensitivities
- Compatible with high-performance LC systems
- Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid
- Tricorn columns are optimized for the polishing step when high purity is required
- PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
- Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
- Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL
Specifications
Specifications
| Buffer | All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid |
| Product Type | Mono S ion exchange column |
| Bed Volume | 1.7 mL |
| Diameter (English) Inner | 0.18 in. |
| Diameter (Metric) Inner | 4.6 mm |
| Quantity | Each |
| Height (English) | 3.93 in. |
| Height (Metric) | 100 mm |
| Flow Rate | 0.05 to 3mL/min. |
| Material | PEEK |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?