missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Maximum resolution achieved from efficiency of small spherical particles, optimally packed
- Particles are coupled to Q or S ion exchangers with excellent sensitivities
- Compatible with high-performance LC systems
- Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid
- Tricorn columns are optimized for the polishing step when high purity is required
- PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
- Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
- Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL
Specifications
Specifications
| Buffer | All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid |
| Product Type | Mono Q ion exchange column |
| Bed Volume | 8mL |
| Diameter (English) Inner | 0.39 in. |
| Diameter (Metric) Inner | 10mm |
| Quantity | Each |
| Height (English) | 3.93 in. |
| Height (Metric) | 100mm |
| Flow Rate | 2 to 6mL/min. |
| Material | Borosilicate Glass |
| Show More |
Safety and Handling
Product Identifier
- Mono Q 10/100 GL
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contain: ethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?