missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MiniBeads™ Columns

Product Code. 10370455 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Mini Q Ion Exchange Column
Mini S Ion Exchange Column
Bed Volume:
0.24 mL
0.8 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Bed Volume unitSize
10370455 Mini Q Ion Exchange Column 0.8 mL Each
10257754 Mini Q Ion Exchange Column 0.24 mL Each
10217804 Mini S Ion Exchange Column 0.24 mL Each
10605395 Mini S Ion Exchange Column 0.8 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10370455 Supplier Cytiva Supplier No. 17517701

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MiniBeads™ prepacked columns are designed for high performance purification and characterization of biomolecules using ion exchange chromatography

  • Small, mono-disperse beads for extremely high resolution purifications. non porous beads contribute to high resolution but give lower capacity than MonoBeads
  • Analytical and micro-preparative applications with choice of both anion and cation exchangers
  • High chemical and physical stability
  • Excellent reproducibility and durability

  • Tricorn high-performance columns (4.6/50 PE) prepacked with MiniBeads
  • PC columns designed for micropurification

Precision column holders may be required for use on high performance chromatography systems such as KTA design. For optimum results, it is important to check the compatibility of the column and the system specifications.

TRUSTED_SUSTAINABILITY

Specifications

Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
Product Type Mini Q Ion Exchange Column
Bed Volume 0.8 mL
Diameter (English) Inner 0.18 in.
Diameter (Metric) Inner 4.6 mm
Quantity Each
Height (English) 1.96 in.
Height (Metric) 50 mm
Flow Rate 0.5 to 2 mL/min.
Includes Valco fittings
Material Polyethere Ketone
Ligand Type O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3
Max. Pressure 180psi
pH Range 3 to 11
Particle Size 3 μm
Support Type Monodisperse Polystyrene/Divinyl Benzene particles
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) High performance purification and characterization of biomolecules by ion exchange chromato graph
Show More Show Less
compliance-icons
Product Identifier
  • Mini Q 4.6/50 PE
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.