missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ S, Capto™ Q and Capto™ DEAE Prepacked IEX Columns

Product Code. 10352780 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
HiScreen Capto DEAE IEX Column
HiScreen Capto Q IEX column
HiScreen Capto S IEX column
Ligand Type:
Diethylaminoethyl
Quaternary Amine
Sulphonate group
pH Range:
2 to 12
2 to 9
4 to 12
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Ligand Type pH Range unitSize
10352780 HiScreen Capto DEAE IEX Column Diethylaminoethyl 2 to 9 Each
10609620 HiScreen Capto Q IEX column Quaternary Amine 2 to 12 Each
10619620 HiScreen Capto S IEX column Sulphonate group 4 to 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10352780 Supplier Cytiva Supplier No. 28926982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for method optimization and parameter screening

  • Fast, efficient, and cost-effective capture and intermediate protein purification
  • Ready-to-use columns for convenient process development optimization
  • Highly cross-linked agarose with dextran surface extender matrix
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

Capto S: Strong cation (sulfonate group) exchange BioProcess™ medium
Capto Q: Strong anion (quartenary amine group) exchange BioProcess medium
Capto DEAE: Weak anion (diethylaminoethyl group) exchange BioProcess medium

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetic Acid, 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type HiScreen Capto DEAE IEX Column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <700 cm/hr.
Material Polypropylene
Ligand Type Diethylaminoethyl
Max. Pressure 3 bar
pH Range 2 to 9
Particle Size 45 to 165μm
Support Type Highly cross-linked Agarose with Dextran surface extender
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Modern large-scale manufacturers for fast, efficient and cost-effective capture and intermediate Protein purification, optimization and parameter screening
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.