missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva CyDye™ DIGE Fluors: Cy3
Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
Brand: Cytiva
Code : GE
Additional Details : Weight : 2.94830kg
Description
- Size- and charge-matched dyes enable co-migration of labeled samples within the gel
- Bright and highly sensitive dyes allow the use of the minimal labeling technique
- Minimal loss of signal during labeling, separation, and scanning
- No change in signal over wide pH range used during first-dimension (IEF) separation
- Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
- Allow the labeling of 50μg protein per labeling reaction
For general 2-D applications where sufficient amounts of sample are available.
Specifications
Solid | |
2 x 5 nM | |
10nM of CyDye DIGE Fluor Cy3 minimal dye | |
-15°C to -30°C |
For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel | |
Amersham CyDye DIGE Fluor Cy3 Minimal Dye | |
For experiments with a limited number of variables, up to 25 labeling | |
Green |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1