missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)

Product Code. 11515095 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Label or Dye:
Cy3
Cy5
Cy5.5
Product Type:
Cy3 Maleimide
Cy5 Maleimide
Cy5.5 Maleimide
Unit Size:
1mg
25mg
5mg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Label or Dye Product Type unitSize
11515095 5 mg Cy3 Cy3 Maleimide 5mg
10479724 1 mg Cy5.5 Cy5.5 Maleimide 1mg
11515085 25 mg Cy5.5 Cy5.5 Maleimide 25mg
11545085 25 mg Cy5 Cy5 Maleimide 25mg
11555085 5 mg Cy5 Cy5 Maleimide 5mg
11525085 25 mg Cy5.5 Cy5.5 Maleimide 5mg
10637475 1 mg Cy3 Cy3 Maleimide 1mg
10248624 1 mg Cy5 Cy5 Maleimide 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11515095 Supplier Cytiva Supplier No. PA13130

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides

The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.

  • Suitable for the labeling of antibodies
  • Example protocols included with each dye pack

Antibody labeling

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For labeling via Thiol groups, use in steady state Fluorescence, FP and FRET applications
Quantity 5 mg
Product Type Cy3 Maleimide
Kit Contents Foil packs, each containing 1, 5 or 25mg dried dye
pH Range 3 to 10
Content And Storage 2°C to 8°C
Label or Dye Cy3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.