missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)

Product Code. 11575045 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Product Type:
Cy2 bis-NHS Ester
Cy3 bis-NHS Ester
Cy5 Bis NHS Ester
Cy5 bis-NHS Ester
Cy5.5 Bis NHS Ester
Cy5.5 bis-NHS Ester
Cy7 Bis NHS Ester
Cy7 bis-NHS Ester
Unit Size:
10mg
50mg
5mg
Pack of 5
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
11575045 50 mg Cy5 Bis NHS Ester 50mg
11585035 5 mg Cy2 bis-NHS Ester Pack of 5
11515045 50 mg Cy3 bis-NHS Ester 50mg
11505045 5 mg Cy3 bis-NHS Ester 5mg
11525045 10 mg Cy3 bis-NHS Ester 10mg
11585045 10 mg Cy5 bis-NHS Ester 10mg
11525055 5 mg Cy5.5 bis-NHS Ester 5mg
11535055 50 mg Cy5.5 Bis NHS Ester 50mg
11555055 5 mg Cy7 bis-NHS Ester 5mg
11565055 10 mg Cy7 Bis NHS Ester 10mg
11565045 5 mg Cy5 bis-NHS Ester 5mg
11545055 10 mg Cy5.5 bis-NHS Ester 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 11575045 Supplier Cytiva Supplier No. PA15002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for general labeling of amine residues

Order Info

Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Used for high-throughput screening (HTS) applications
Quantity 50 mg
Product Type Cy5 Bis NHS Ester
Kit Contents Foil packs, each containing 5, 10 or 50mg dried dye
Molecular Weight (g/mol) 975.15
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Color Blue
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Cy5 (Bis NHS ester)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.