missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN2, p Thr899 Antibody (SR1336), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | GCN2 |
|---|---|
| Clone | SR1336 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641885
|
Novus Biologicals
NBP3-21513-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611114
|
Novus Biologicals
NBP3-21513-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCN2 Monoclonal antibody specifically detects GCN2 in Human samples. It is validated for Western BlotSpecifications
| GCN2 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1 | |
| A synthesized peptide derived from human phospho-GCN2 [Thr899] (Uniprot #: Q9P2K8) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1336 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 440275 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title