missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN2, p Thr899 Antibody (SR1336), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21513-100ul
This item is not returnable.
View return policy
Description
GCN2 Monoclonal antibody specifically detects GCN2 in Human samples. It is validated for Western Blot
Specifications
| GCN2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1336 | |
| Western Blot 1:500-1:2000 | |
| eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1 | |
| A synthesized peptide derived from human phospho-GCN2 [Thr899] (Uniprot #: Q9P2K8) | |
| 100 μg | |
| Phospho Specific, Protein Kinase | |
| 440275 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction