missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | GBX1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
GBX1 Polyclonal specifically detects GBX1 in Human samples. It is validated for Western Blot.Specifications
| GBX1 | |
| Polyclonal | |
| Rabbit | |
| NP_001092304 | |
| 2636 | |
| IgG | |
| 40 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| gastrulation brain homeo box 1, gastrulation brain homeobox 1, homeobox protein GBX-1 | |
| GBX1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title