missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91420
This item is not returnable.
View return policy
Description
GBX1 Polyclonal specifically detects GBX1 in Human samples. It is validated for Western Blot.
Specifications
| GBX1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| gastrulation brain homeo box 1, gastrulation brain homeobox 1, homeobox protein GBX-1 | |
| Rabbit | |
| 40 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Fruit fly: 100%; Medaka fish: 100%; Green puffer: 100%; Body louse: 100%; Red flour beetle: 100%; African malaria mosquito: 92%; Western clawed frog: 92%; Florida lancelet: 92%; Yellowfever mosquito: 92%; Common carp: 92%; Mosquito: 92%; Xenopus: 92%; Southern house mosquito: 92%; Common lancelet: 91%; Budgerigar: 85%; Bovine: 85%; Canine: 85%; Dumeril's clam worm: 85%; Bobwhite quail: 85%; Bornean orangutan: 85%; Chimpanzee: 85%; Pig-tailed macaque: 85%; Common woolly monkey: 85%; Black-handed spider monkey: 85%; Pygmy chimpanzee: 85%; Western Gorilla: 78%; Gibbula varia: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_001092304 | |
| GBX1 | |
| The immunogen is a synthetic peptide directed towards the middle region of human GBX1. Peptide Sequence AKWKRIKAGNVSSRSGEPVRNPKIVVPIPVHVNRFAVRSQHQQMEQGARP. The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 2636 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction