missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA2/GATA3 Recombinant Rabbit Monoclonal Antibody (23GB5730), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | GATA2/GATA3 |
|---|---|
| Clone | 23GB5730 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088124
![]() |
Invitrogen™
MA553920 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088803
|
Invitrogen™
MA553921 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GATA2/GATA3 Recombinant Monoclonal antibody specifically detects GATA2/GATA3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GATA2/GATA3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O09100, P23769, P23771, P23772, Q924Y4 | |
| 14461, 14462, 25159, 2624, 2625, 85471 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5730 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| DCML; EMBL:AAK51128.1}; endothelial transcription factor GATA-2; GATA binding protein 2; GATA binding protein 3; Gata2; Gata-2; gata2 {ECO:0000312; Gata3; Gata-3; GATA-binding factor 3; GATA-binding protein 2; GATA-binding protein 3; GATA-bindning protein 2; HDR; HDRS; IMD21; jal; MGC2346; MGC5199; MGC5445; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3 | |
| A synthesized peptide derived from human GATA2/3 (1-250AA). | |
| Primary | |
| -20°C | |
| GATA2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title