missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA2/GATA3 Recombinant Rabbit Monoclonal Antibody (23GB5730), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553921
This item is not returnable.
View return policy
Description
The genes for all 4 subunits of the T-cell antigen receptor (alpha, beta, gamma and delta) are controlled by distinct enhancers and their enhancer-binding proteins. Marine and Winoto (1991) identified a common TCR regulatory element by demonstrating binding of the enhancer-binding protein GATA3 to the enhancer elements of all 4 TCR genes. GATA3 had been shown in the chicken to be an enhancer-binding protein containing a zinc finger domain. GATA3 mRNA was demonstrated by Northern blot analysis in T cells but not in B cells or macrophages. GATA3 is abundantly expressed in the T-lymphocyte lineage and is thought to participate in T-cell receptor gene activation through binding to enhancers. Labastie et al. (1994) cloned the human gene and the 5-prime end of the mouse gene. The human gene comprises 6 exons distributed over 17 kb of DNA. Its 2 zinc fingers are encoded by 2 separate exons highly conserved with those of GATA1, but no other structural homologies between the 2 genes could be found.
Specifications
| GATA2/GATA3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GATA2 | |
| DCML; EMBL:AAK51128.1}; endothelial transcription factor GATA-2; GATA binding protein 2; GATA binding protein 3; Gata2; Gata-2; gata2 {ECO:0000312; Gata3; Gata-3; GATA-binding factor 3; GATA-binding protein 2; GATA-binding protein 3; GATA-bindning protein 2; HDR; HDRS; IMD21; jal; MGC2346; MGC5199; MGC5445; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14461, 14462, 25159, 2624, 2625, 85471 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5730 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O09100, P23769, P23771, P23772, Q924Y4 | |
| GATA2, GATA3 | |
| A synthesized peptide derived from human GATA2/3 (1-250AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction