missing translation for 'onlineSavingsMsg'
Learn More

GATA-4 Antibody (Evan) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody

£278.00 - £400.00

Specifications

Antigen GATA-4
Clone Evan
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18256721
View Documents
Novus Biologicals
NBP1-43112-0.025MG
0.025 mg
£278.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18271042
View Documents
Novus Biologicals
NBP1-43112-0.1MG
0.1 mg
£400.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

GATA-4
0.5 mg/mL
Western Blot
Unconjugated
RUO
Human, Mouse
GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
GATA4
IgG2a κ
Affinity Purified
Evan
Western Blot 1:100-1:2000
Monoclonal
Rat
Mesenchymal Stem Cell Markers, Stem Cell Markers
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
2626
The immunogen for this antibody was Gata4.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.