missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-4 Antibody (Evan) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43112-0.1MG
This item is not returnable.
View return policy
Description
GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| GATA-4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000 | |
| GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a κ |
| Western Blot | |
| Evan | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| GATA4 | |
| The immunogen for this antibody was Gata4. | |
| 0.1 mg | |
| Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2626 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction