Learn More
GATA-3 Recombinant Rat Monoclonal Antibody (TWAJ), Invitrogen™
Rat Recombinant Monoclonal Antibody
£225.00 - £450.00
Specifications
| Antigen | GATA-3 |
|---|---|
| Clone | TWAJ |
| Concentration | 1.0 mg/mL |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30130997
|
Invitrogen
740023T20UG |
20 μL |
£225.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30131054
|
Invitrogen
740023T |
100 μL |
£450.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
GATA-3 Recombinant Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| GATA-3 | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3 | |
| IgG2b κ | |
| Affinity Chromatography | |
| Antibody |
| TWAJ | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P23771 | |
| 2625 | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GATA3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.