Learn More
GATA-3 Recombinant Rat Monoclonal Antibody (TWAJ), Invitrogen™
Rat Recombinant Monoclonal Antibody
Brand: Invitrogen 740023T20UG
Description
Recombinant rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
GATA-3 Recombinant Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| GATA-3 | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P23771 | |
| GATA3 | |
| Affinity Chromatography | |
| RUO | |
| 2625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| TWAJ | |
| Unconjugated | |
| GATA3 | |
| EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3 | |
| Rat | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.