missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrokine 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | Gastrokine 1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.1-0.2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159641
|
Novus Biologicals
NBP2-24766SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18146105
|
Novus Biologicals
NBP2-24766 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gastrokine 1 Polyclonal specifically detects Gastrokine 1 in Human, Porcine, Equine, Primate samples. It is validated for Western Blot.Specifications
| Gastrokine 1 | |
| Western Blot 0.1-0.2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Pig, Equine, Primate | |
| 18 kDa antrum mucosa protein, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, Protein CA11 | |
| GKN1 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and dog, 94% homologous in mouse and cow, 88% homologous in rat and 77% homologous in chicken. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 56287 | |
| The portions of amino acids range between 100-150 of human GKN1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title