missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrokine 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-24766
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Gastrokine 1 Polyclonal specifically detects Gastrokine 1 in Human, Porcine, Equine, Primate samples. It is validated for Western Blot.
Specifica
| Gastrokine 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| GKN1 | |
| The portions of amino acids range between 100-150 of human GKN1 was used as the immunogen. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 56287 | |
| Human, Pig, Equine, Primate | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.1-0.2 ug/ml | |
| 18 kDa antrum mucosa protein, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, Protein CA11 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and dog, 94% homologous in mouse and cow, 88% homologous in rat and 77% homologous in chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction