missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastric Lipase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£165.00 - £367.00
Specifications
| Antigen | Gastric Lipase |
|---|---|
| Concentration | 0.9 mg/mL |
| Dilution | Western Blot 2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290311
|
Novus Biologicals
NB110-60930SS |
0.025 mL |
£165.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18278570
|
Novus Biologicals
NB110-60930 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gastric Lipase Polyclonal specifically detects Gastric Lipase in Human, Mouse samples. It is validated for Western Blot.Specifications
| Gastric Lipase | |
| Western Blot 2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Autophagy, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Endothelial Cell Markers, Glia Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271 | |
| LIPF | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen sequence has 87% homology to canine. |
| 0.9 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P07098 | |
| 8513 | |
| A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot# P07098] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 45 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title