missing translation for 'onlineSavingsMsg'
Learn More

Gastric Lipase Antibody - BSA Free, Novus Biologicals™

Product Code. 18290311 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18290311 0.025 mL 25µL
18278570 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18290311 Supplier Novus Biologicals Supplier No. NB11060930SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gastric Lipase Polyclonal specifically detects Gastric Lipase in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gastric Lipase
Applications Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. P07098
Gene Alias EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271
Gene Symbols LIPF
Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot# P07098]
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Endothelial Cell Markers, Glia Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8513
Test Specificity Human and mouse. Immunogen sequence has 87% homology to canine.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.