Learn More
Ekastu Safety™ Gastight Goggle
Product Code. 10702562
Change view
Click to view available options
UV Protection:
Yes
Unit Size:
Each
Product Code. UV Protection unitSize
10702562 Yes Each
1 options
Product Code. 10702562 Supplier Ekastu Safety™ Supplier No. 477231
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Gas tight and dustproof full-view goggles resistant to chemicals. EKASTU Safety™ Gas tight Goggles include anti-fogging lenses with 99.99% UV protection and innovative UV 380 technology.
  • DIN EN 166 1 5 – SN
  • Full view goggles resistant to chemicals
  • Gas tight and dustproof
  • Made of rubber which is kind to the skin
  • With clear three-layer sandwich lenses
  • Anti-fogging
  • Large field of vision
  • Quick and easy lens replacement
  • Comfortable, pressure-free fit
  • 99.99% UV protection with innovative UV 380 technology
Specifications
Certifications/Compliance DIN EN 166 1 5 – SN
Lens Coating Anti-fog
Product Type Gastight Goggle
For Use With (Application) Gas and dust applications, chemical resistant
Frame Color Black
No. per Pack 1 Ea.
Material Rubber
UV Protection Yes