missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gassner Medium

Product Code. 10291443
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10291443 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10291443 Supplier Thermo Scientific™ Supplier No. PO5021A
  • ONLINE EXCLUSIVE PRICE
    £12.10
    £11.46 / Pack of 10
    Save £0.64
    5% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect and isolate Enterobacteriaceae from food samples with this ready-to-use, selective medium.

Detect and isolate pathogenic Enterobacteriaceae from food samples with Thermo Scientific™ Oxoid™ Gassner Medium. It is a modification of the original Gassner (water blue metachrome yellow lactose) medium1. The medium contains metachrome yellow, which inhibits the Gram-positive accompanying flora. Lactose-digesting microorganisms such as Escherichia coli or Klebsiella species form blue colonies with simultaneous blue colouring of the surrounding agar. Lactose-negative microorganisms, such as Salmonella, Serratia or Proteus species, form yellow colonies with a yellow background.

Use Gassner Medium for selective detection and isolation of Enterobacteriaceae from food and other samples.

  • Selective: presence of metachrome yellow inhibits non-target Gram-positive flora.
  • Easy-to-read: Lactose positive organisms form blue colonies while lactose negative organisms form yellow colonies.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Olive Green
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.