Learn More
Macherey-Nagel™ Gasket
Product Code. 10457241 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
Lid and 24 Columns
Unit Size:
Pack of 2
Product Code. For Use With (Equipment) Unit Size
10457241 Lid and 24 Columns Pack of 2
1 options
Product Code. 10457241 Supplier Macherey-Nagel™ Supplier No. 730178
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with vacuum manifolds
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.
Specifications
For Use With (Equipment) Lid and 24 Columns
Type Gasket
Quantity 2 Pack