missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GAS2 Recombinant Rabbit Monoclonal Antibody (24GB3195)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen GAS2
Clone 24GB3195
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275303
GREENER_CHOICE
View Documents
Invitrogen™
MA556932
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275025
View Documents
Invitrogen™
MA556933
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GAS2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Gas2 is a 313 amino acid protein encoded by the human gene GAS2. Gas2 is thought to play a role in apoptosis by acting as a cell death substrate for caspases. Gas2, a component of the microfilament system, is cleaved by a caspase (caspase-3 and caspase-7) at Asparagine 278 during apoptosis. The cleaved form resulting from this dramatically induces the rearrangement of the Actin cytoskeleton and causes potent changes in the shape of the affected cells. Gas2 is believed to also be involved in the membrane ruffling process. During the G0-G1 transition phase Gas2 can be found phosphorylated on its serine residues. Gas2 is a cytoskeleton and peripheral membrane protein that co-localizes with Actin fibers at the cell border and along the stress fibers in growth-arrested fibroblasts. Gas2 is mainly membrane-associated but when hyperphosphorylated it will accumulate at membrane ruffles. Gas2 is specifically expressed at growth arrest and is ubiquitously expressed with highest levels found in liver, lung and kidney.
Specifications

Specifications

GAS2
-20°C
Recombinant Monoclonal
Liquid
IgG
Mouse
GAS2
GAS 2; Gas2; Gas-2; growth arrest specific 2; growth arrest-specific 2; growth arrest-specific protein 2; RGD1563167
GAS2
Primary
Affinity chromatography
24GB3195
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P11862
14453
A synthesized peptide derived from human GAS2 (aa 1-56).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.