missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43, p Ser41 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £372.00
Specifications
| Antigen | GAP-43 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632397
|
Novus Biologicals
NBP3-05419-100ul |
100 μg |
£372.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609106
|
Novus Biologicals
NBP3-05419-20ul |
20 μg |
£159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAP-43 Polyclonal antibody specifically detects GAP-43 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GAP-43 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| PBS with 50% glycerol, pH7.3. | |
| 2596 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
| A synthetic phosphorylated peptide around S41 of human GAP-43 (NP_002036.1). QASFR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title