missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43, p Ser41 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05419-20ul
This item is not returnable.
View return policy
Description
GAP-43 Polyclonal antibody specifically detects GAP-43 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| GAP-43 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
| A synthetic phosphorylated peptide around S41 of human GAP-43 (NP_002036.1). QASFR | |
| 20 μg | |
| Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 2596 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction