missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAMT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54399
This item is not returnable.
View return policy
Description
GAMT Polyclonal specifically detects GAMT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GAMT | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.1.1.2, guanidinoacetate N-methyltransferase, PIG2, TP53I2 | |
| Rabbit | |
| 29 kDa | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 2593 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| A8K0A0 | |
| GAMT | |
| Synthetic peptides corresponding to GAMT(guanidinoacetate N-methyltransferase) The peptide sequence was selected from the middle region of GAMT. Peptide sequence PGEGPFLTPWVGWTVLVHLEIKVLCLAQWLPGAVAQVYNPSTVEGRGGQI. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction