missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Synuclein Antibody (1H10D2), Novus Biologicals™
Mouse Monoclonal Antibody
£139.00
Specifications
| Antigen | gamma-Synuclein |
|---|---|
| Clone | 1H10D2 |
| Dilution | Western Blot 1: 500-1: 2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| gamma-Synuclein | |
| Western Blot 1: 500-1: 2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1) | |
| SNCG | |
| IgG1 | |
| Unpurified | |
| SNCG (1H10D2) |
| 1H10D2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Cellular Markers, Mitotic Regulators, Neuroscience | |
| PBS with 0.03% Sodium Azide | |
| 6623 | |
| Ni-NTA purified truncated recombinant gamma-Synuclein Antibody (1H10D2) expressed in E. Coli strain BL21 (DE3) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title