missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Synuclein Antibody (1H10D2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60497-0.025ml
This item is not returnable.
View return policy
Description
gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| gamma-Synuclein | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.03% Sodium Azide | |
| SNCG | |
| Ni-NTA purified truncated recombinant gamma-Synuclein Antibody (1H10D2) expressed in E. Coli strain BL21 (DE3) | |
| 0.025 mL | |
| Alzheimers Research, Cellular Markers, Mitotic Regulators, Neuroscience | |
| 6623 | |
| Human | |
| Ascites |
| Western Blot | |
| 1H10D2 | |
| Western Blot 1: 500-1: 2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1) | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| SNCG (1H10D2) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction