missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ gamma-glutamyl hydrolase Antibody (SR4337), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | gamma-glutamyl hydrolase |
|---|---|
| Clone | SR4337 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041650
|
Novus Biologicals™
NBP3-27850-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041608
|
Novus Biologicals™
NBP3-27850-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma-glutamyl hydrolase Monoclonal antibody specifically detects gamma-glutamyl hydrolase in Human samples. It is validated for Western BlotSpecifications
| gamma-glutamyl hydrolase | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Conjugase, EC 3.4.19.9, gamma-glutamyl hydrolase, gamma-glutamyl hydrolase (conjugase, folylpolygammaglutamyl hydrolase), Gamma-Glu-X carboxypeptidase, GH | |
| A synthesized peptide derived from human gamma-glutamyl hydrolase (Uniprot #: Q92820) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR4337 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 8836 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title