missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ gamma-glutamyl hydrolase Antibody (SR4337), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27850-100ul
This item is not returnable.
View return policy
Description
gamma-glutamyl hydrolase Monoclonal antibody specifically detects gamma-glutamyl hydrolase in Human samples. It is validated for Western Blot
Specifications
| gamma-glutamyl hydrolase | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR4337 | |
| Western Blot 1:500-1:2000 | |
| Conjugase, EC 3.4.19.9, gamma-glutamyl hydrolase, gamma-glutamyl hydrolase (conjugase, folylpolygammaglutamyl hydrolase), Gamma-Glu-X carboxypeptidase, GH | |
| A synthesized peptide derived from human gamma-glutamyl hydrolase (Uniprot #: Q92820) | |
| 100 μL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 8836 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction