missing translation for 'onlineSavingsMsg'
Learn More

gamma-Glu-Cys Trifluoroacetic Acid Salt (~90%), TRC

Product Code. 30240695
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240695 5 mg 5mg
30240651 50 mg 50mg
30240529 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240695 Supplier Toronto Research Chemicals Supplier No. TRCG5971055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 283159-88-6
Molecular Formula C8H14N2O5S • x(C2HF3O2)
Molecular Weight (g/mol) 250.2711402
Synonym L-Cysteine, L-γ-glutamyl-2,2,2-Trifluoroacetic Acid Salt, L-Cysteine, L-γ-glutamyl- Trifluoroacetic Acid Salt;
SMILES OC([C@H](CS)NC(CC[C@H](N)C(O)=O)=O)=O.OC(C(F)(F)F)=O.[F,Cl,Br,I]

Specifications

InChI Formula InChI=1S/C8H14N2O5S.C2HF3O2/c9-4(7(12)13)1-2-6(11)10-5(3-16)8(14)15;3-2(4,5)1(6)7/h4-5,16H,1-3,9H2,(H,10,11)(H,12,13)(H,14,15);(H,6,7)/t4-,5-;/m0./s1
Quantity 5 mg
Shipping Condition Room Temperature
Chemical Name or Material gamma-Glu-Cys Trifluoroacetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.