CAS RN 283159-88-6
CAS RN 283159-88-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
gamma-Glu-Cys Trifluoroacetic Acid Salt (~90%), TRC
CAS: 283159-88-6 Molecular Formula: C8H14N2O5S • x(C2HF3O2) Molecular Weight (g/mol): 250.2711402 Synonym: L-Cysteine, L-γ-glutamyl-2,2,2-Trifluoroacetic Acid Salt,L-Cysteine, L-γ-glutamyl- Trifluoroacetic Acid Salt; SMILES: OC([C@H](CS)NC(CC[C@H](N)C(O)=O)=O)=O.OC(C(F)(F)F)=O.[F,Cl,Br,I]