missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Catenin Rabbit anti-Human, Rat, Bovine, Canine, Chicken, Feline, Clone: CTNG/2155R, Novus Biologicals™
Rabbit Monoclonal Antibody
£236.00
Specifications
| Antigen | gamma Catenin |
|---|---|
| Clone | CTNG/2155R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
Description
gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Rat, Bovine, Canine, Chicken, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| gamma Catenin | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 3728 | |
| IgG | |
| Protein A or G purified |
| CTNG/2155R | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Bovine, Canine, Chicken, Feline | |
| catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
| Recombinant, human full-length gamma Catenin (CTNG) protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title