missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Catenin Rabbit anti-Human, Rat, Bovine, Canine, Chicken, Feline, Clone: CTNG/2155R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07594-20UG
This item is not returnable.
View return policy
Description
gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Rat, Bovine, Canine, Chicken, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| gamma Catenin | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
| catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
| Recombinant, human full-length gamma Catenin (CTNG) protein | |
| 20 μg | |
| Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
| 3728 | |
| Store at 4°C. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| CTNG/2155R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Bovine, Canine, Chicken, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction