missing translation for 'onlineSavingsMsg'
Learn More

(-)-Gallocatechin Gallate, TRC

Product Code. 30132981
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132981 25 mg 25mg
30007239 10 mg 10mg
30010492 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132981 Supplier Toronto Research Chemicals Supplier No. TRCG18898525MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4233-96-9
Molecular Formula C22 H18 O11
Molecular Weight (g/mol) 458.37
Synonym Benzoic acid, 3,4,5-trihydroxy-, (2S,3R)-3,4-dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3-yl ester, Benzoic acid, 3,4,5-trihydroxy-, 3,4-dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3-yl ester, (2S-trans)-, Gallocatechol, 3-gallate, (-)- (8CI), Gallic acid, ester with gallocatechol, (-) (8CI), (-)-Gallocatechin 3-O-gallate, (-)-Gallocatechin 3-gallate, (-)-Gallocatechin gallate, (-)-Gallocatechol gallate, L-GCG, NVP-XAA 225
IUPAC Name [(2S,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@@H](Oc2c1)c4cc(O)c(O)c(O)c4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21+/m1/s1
Formula Weight 458.0849
Chemical Name or Material (-)-Gallocatechin Gallate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.