missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanin Antibody (4B3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £449.00
Specifications
| Antigen | Galanin |
|---|---|
| Clone | 4B3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692389
|
Novus Biologicals
NBP3-09010-0.025mg |
0.025 mg |
£216.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688167
|
Novus Biologicals
NBP3-09010-0.2mg |
0.2 mg |
£449.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galanin Monoclonal antibody specifically detects Galanin in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| Galanin | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167 | |
| C-terminal fusions of KLH to human galanin, | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 4B3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Neutralization | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| PBS | |
| 51083 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title