missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanin Antibody (4B3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09010-0.2mg
This item is not returnable.
View return policy
Description
Galanin Monoclonal antibody specifically detects Galanin in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Neutralization
Specifications
| Galanin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Neutralization | |
| 4B3 | |
| Unconjugated | |
| GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167 | |
| C-terminal fusions of KLH to human galanin, | |
| 0.2 mg | |
| Neuroscience | |
| 51083 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction