missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD153/CHOP Antibody (9C8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 34 publications
£193.00 - £426.00
Specifications
| Antigen | GADD153/CHOP |
|---|---|
| Clone | 9C8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Simple Western 1:250, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP) |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18777573
|
Novus Biologicals
NB600-1335SS |
0.025 mL |
£193.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18719263
|
Novus Biologicals
NB600-1335 |
0.1 mL |
£426.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GADD153/CHOP Monoclonal specifically detects GADD153/CHOP in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP), Knockdown Validated.Specifications
| GADD153/CHOP | |
| 1 mg/mL | |
| ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Cycle and Replication, DNA Repair, Hypoxia, Lipid and Metabolism, Neuroscience, Unfolded Protein Response | |
| P35638, P35639 | |
| 1649 | |
| Full length mouse CHOP/GADD153 [Swiss-Prot# P35639] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 9C8 | |
| Western Blot, Simple Western 1:250, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| C/EBP-homologous protein, C/EBP-homologous protein 10, CCAAT/enhancer-binding protein homologous protein, CHOP-10, CHOP10 CEBPZ, CHOPC/EBP zeta, DDIT-3, DNA damage-inducible transcript 3 protein, DNA-damage-inducible transcript 3, GADD153, Growth arrest and DNA damage-inducible protein GADD153 | |
| DDIT3 | |
| IgG2b κ | |
| Protein A purified | |
| 19 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title