missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD153/CHOP Antibody (9C8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 34 publications
Brand: Novus Biologicals NB600-1335
This item is not returnable.
View return policy
Description
GADD153/CHOP Monoclonal specifically detects GADD153/CHOP in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP), Knockdown Validated.
Specifications
| GADD153/CHOP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Simple Western 1:250, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP) | |
| C/EBP-homologous protein, C/EBP-homologous protein 10, CCAAT/enhancer-binding protein homologous protein, CHOP-10, CHOP10 CEBPZ, CHOPC/EBP zeta, DDIT-3, DNA damage-inducible transcript 3 protein, DNA-damage-inducible transcript 3, GADD153, Growth arrest and DNA damage-inducible protein GADD153 | |
| Mouse | |
| 19 kDa | |
| 0.1 mL | |
| Apoptosis, Cell Cycle and Replication, DNA Repair, Hypoxia, Lipid and Metabolism, Neuroscience, Unfolded Protein Response | |
| 1649 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 9C8 | |
| Unconjugated | |
| P35638, P35639 | |
| DDIT3 | |
| Full length mouse CHOP/GADD153 [Swiss-Prot# P35639] | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction