missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GABRG2 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16852594
|
Proteintech
14104-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16842594
|
Proteintech
14104-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.Specifications
| GABRG2 | |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P18507, P18508, P22723 | |
| 14406, 2566, 29709 | |
| GABRG2 Fusion Protein Ag5237 | |
| Primary | |
| -20°C | |
| GABRG2 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CAE2, ECA2, GABA A receptor gamma 2, GABAA receptor gamma 2, GABRG2, GEFSP3 | |
| Gabrg2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title