Learn More
GABRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14104-1-AP-20UL
 View more versions of this product
Product Code. 16852594
  • £135.00 / 20µL
Description
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.
Specifications
Specifications
GABRG2
Polyclonal
Unconjugated
GABRG2
CAE2, ECA2, GABA A receptor gamma 2, GABAA receptor gamma 2, GABRG2, GEFSP3
Rabbit
Antigen Affinity Chromatography
RUO
14406, 2566, 29709
-20°C
Liquid
Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18507, P18508, P22723
Gabrg2
GABRG2 Fusion Protein Ag5237
20 μL
Primary
Rat, Human, Mouse