missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRA4 Monoclonal Antibody (K56008_4C7), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | GABRA4 |
|---|---|
| Clone | K56008_4C7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: AKRKTSKPPQ EVPAAPVQRE KHPEAPLQNT NANLNMRKRT NALVHSESDV GNRTEVGNHS SKSSTVVQES SKGTPRSYLA SSPNPFSRAN AAETISAARA LPSASPTSIR TGYMPRKASV GSASTRHVFG SRLQRIKTTV NTIGATGKLS ATPPPSAPPP SGSGTSKIDK
GABRA4 Monoclonal antibody specifically detects GABRA4 in Human samples. It is validated for Western BlotSpecifications
| GABRA4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| GABA(A) receptor; GABA(A) receptor subunit alpha-4; GABA(A) receptor, alpha 4; GABA-A receptor alpha-4 subunit; GABAARalpha4; Gabra4; Gabra-4; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 4; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 4; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 4; gamma-aminobutyric acid A receptor alpha 4; gamma-aminobutyric acid receptor subunit alpha-4; gamma-aminobutyric acid type A receptor alpha4 subunit | |
| Recombinant human GABRA4 (immunogen range is 351-520). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GABRA4 |
| K56008_4C7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P48169 | |
| 2557 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title