missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRA4 Monoclonal Antibody (K56008_4C7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555744
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: AKRKTSKPPQ EVPAAPVQRE KHPEAPLQNT NANLNMRKRT NALVHSESDV GNRTEVGNHS SKSSTVVQES SKGTPRSYLA SSPNPFSRAN AAETISAARA LPSASPTSIR TGYMPRKASV GSASTRHVFG SRLQRIKTTV NTIGATGKLS ATPPPSAPPP SGSGTSKIDK
Gamma-aminobutyric acid is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. This gene encodes subunit alpha-4, which is involved in the etiology of autism and eventually increases autism risk through interaction with another subunit, gamma-aminobutyric acid receptor beta-1. Alternatively spliced transcript variants encoding different isoforms have been found in this gene.
Specifications
| GABRA4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P48169 | |
| GABRA4 | |
| Recombinant human GABRA4 (immunogen range is 351-520). | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K56008_4C7 | |
| Unconjugated | |
| GABRA4 | |
| GABA(A) receptor; GABA(A) receptor subunit alpha-4; GABA(A) receptor, alpha 4; GABA-A receptor alpha-4 subunit; GABAARalpha4; Gabra4; Gabra-4; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 4; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 4; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 4; gamma-aminobutyric acid A receptor alpha 4; gamma-aminobutyric acid receptor subunit alpha-4; gamma-aminobutyric acid type A receptor alpha4 subunit | |
| Mouse | |
| Protein A | |
| RUO | |
| 2557 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction