missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABPB1 Recombinant Rabbit Monoclonal Antibody (7B5F5)
Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | GABPB1 |
|---|---|
| Clone | 7B5F5 |
| Concentration | 1.78 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284012
|
Invitrogen™
MA557941 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283338
|
Invitrogen™
MA557942 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: TSGIGQPIIV TMPDGQQVLT VPATDIAEET VISEEPPAKR QCIEIIENRV ESAEIEEREA LQKQLDEANR EAQKYRQQLL KKEQEAEAYR QKLEAMTRLQ TNKEAV.
Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.Specifications
| GABPB1 | |
| 1.78 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q06547 | |
| 2553 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-395 of human GABPB1 (NP_005245.2). | |
| Antibody |
| 7B5F5 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| GABPB1 | |
| BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4Tf1B; GA binding protein subunit beta1; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor, beta subunit 1; GA repeat binding protein, beta 1; GA-binding protein subunit beta-1; Gabp beta1; GABP subunit beta-1; GABP subunit beta-2; GABPB; Gabpb1; GABPB-1; GABPB1-1; GABPB1-2; GABPB2; GABPB-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1560391; transcription factor E4TF1-47; Transcription factor E4TF1-53 | |
| GABPB1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title