missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GABPB1 Recombinant Rabbit Monoclonal Antibody (7B5F5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557941

 View more versions of this product

Product Code. 30284012

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TSGIGQPIIV TMPDGQQVLT VPATDIAEET VISEEPPAKR QCIEIIENRV ESAEIEEREA LQKQLDEANR EAQKYRQQLL KKEQEAEAYR QKLEAMTRLQ TNKEAV.

Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GABPB1
Recombinant Monoclonal
1.78 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q06547
GABPB1
Recombinant fusion protein containing a sequence corresponding to amino acids 290-395 of human GABPB1 (NP_005245.2).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
7B5F5
Unconjugated
GABPB1
BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4Tf1B; GA binding protein subunit beta1; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor, beta subunit 1; GA repeat binding protein, beta 1; GA-binding protein subunit beta-1; Gabp beta1; GABP subunit beta-1; GABP subunit beta-2; GABPB; Gabpb1; GABPB-1; GABPB1-1; GABPB1-2; GABPB2; GABPB-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1560391; transcription factor E4TF1-47; Transcription factor E4TF1-53
Rabbit
Protein A
RUO
2553
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.